Merck
  • Development of an enzyme-linked immunosorbent assay for quantitative determination of quizalofop-p-ethyl.
Development of an enzyme-linked immunosorbent assay for quantitative determination of quizalofop-p-ethyl.
Journal of agricultural and food chemistry (2006-11-09)
Deyi Zeng, Haiyan Shi, Bo Li, Minghua Wang, Baoan Song
ABSTRACT
Accurate quantification of quizalofop-p-ethyl is essential for it may do harm to humans and animals through both water and food. Currently, detection of quizalofop-p-ethyl mainly relies on methods such as gas chromatography, high performance liquid chromatography, and gas chromatography-mass spectrometry. Although these techniques are reliable, they are relatively expensive and time-consuming because of multistep sample cleanup. To address this, we developed a competitive indirect enzyme-linked immunosorbent assay (ciELISA) with a polyclonal antibody against quizalofop-p-ethyl that was generated in our lab. The IC(50) of detection was 0.03495 microg/mL, and the lowest detection limit reached 0.00192 microg/mL. Furthermore, the method had high specificity for it did not cross-react with other structure-related compounds. When water and soil samples that were fortified with quizalofop-p-ethyl were analyzed by this ELISA, recoveries were in the range of 89-110% from water and 81-108% from soil. Good correlations between this immunoassay and gas chromatography data were obtained for residues of quizalofop-p-ethyl in water and soil. Our data indicate that this method is a convenient analytical technique for monitoring quizalofop-p-ethyl in waters without extraction and the extra cleanup step and in soil without the cleanup step.
Product Description
Supelco
Quizalofop-ethyl, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Quizalofop-P-ethyl, PESTANAL®, analytical standard
Pricing and availability is not currently available.