Merck
  • Dynamic synthesis of a macrocycle containing a porphyrin and an electron donor.
Dynamic synthesis of a macrocycle containing a porphyrin and an electron donor.
Chemical communications (Cambridge, England) (2005-03-30)
Amy L Kieran, Sofia I Pascu, Thibaut Jarrosson, Maxwell J Gunter, Jeremy K M Sanders
ABSTRACT
New macrocycles incorporating a porphyrin and a [small pi] electron-rich aromatic were prepared from a dynamic disulfide library. The outcome could be influenced by use of templates.
Sigma-Aldrich
1,3-Dihydroxynaphthalene, ≥99%, crystalline
Pricing and availability is not currently available.
Supelco
1,3-Dihydroxynaphthalene, for spectrophotometric det. of glucuronic acid according to Tollens, ≥97.0%
Pricing and availability is not currently available.