Merck
  • Novel mutual pro-drugs of 2',3'-dideoxyinosine with 3-octadecyloxy-propane-1,2-diol by straightforward enzymatic regioselective synthesis in acetone.
Novel mutual pro-drugs of 2',3'-dideoxyinosine with 3-octadecyloxy-propane-1,2-diol by straightforward enzymatic regioselective synthesis in acetone.
Biotechnology letters (2005-02-11)
Xiao-Feng Sun, Qi Wu, Na Wang, Ying Cai, Xian-Fu Lin
ABSTRACT
Novel mutual pro-drugs in which 2',3'-dideoxyinosine anti-HIV (human immunodeficiency virus) drug and 3-octadecyloxy-propane-1,2-diol anti-inflammatory drug were attached to the same molecule via different biodegradable linkages, were synthesized through two-step enzymatic transesterification by a commercial lipase in acetone.
Sigma-Aldrich
Batyl alcohol, 99%