Merck
  • Synthesis and structure-activity study of fungicidal anilinopyrimidines leading to mepanipyrim (KIF-3535) as an anti-Botrytis agent.
Synthesis and structure-activity study of fungicidal anilinopyrimidines leading to mepanipyrim (KIF-3535) as an anti-Botrytis agent.
Pest management science (2004-05-04)
Toshihiro Nagata, Katsumi Masuda, Shinichiro Maeno, Ichiro Miura
ABSTRACT
A series of 2-anilinopyrimidines was prepared and their fungicidal activities against Botrytis cinerea Pers were examined. The activity fell sharply with any substitution on the anilinobenzene ring. Substituents at the 5-position of the pyrimidine ring greatly reduced the activity. Substituents such as chloro, methoxy, methylamino, methyl or 1-propynyl were well tolerated at the 4- and 6-positions of the pyrimidine ring. Among these substituents, the combination of methyl and 1-propynyl groups was the most favourable. 2-Anilino-4-methyl-6-(1-propynyl)pyrimidine (KIF-3535), which showed excellent activity and no significant phytotoxicity, was finally selected for development and has been given the common name mepanipyrim.
Product Description
Supelco
Mepanipyrim, PESTANAL®, analytical standard