Merck
  • Use of 13C as an indirect tag in 15N specifically labeled nucleosides. Syntheses of [8-13C-1,7,NH2-15N3]adenosine, -guanosine, and their deoxy analogues.
Use of 13C as an indirect tag in 15N specifically labeled nucleosides. Syntheses of [8-13C-1,7,NH2-15N3]adenosine, -guanosine, and their deoxy analogues.
The Journal of organic chemistry (2003-10-25)
Anthony J Shallop, Barbara L Gaffney, Roger A Jones
ABSTRACT
We have previously reported the use of a 13C tag at the C2 of 15N-multilabeled purine nucleosides to distinguish the adjacent-labeled 15N atoms from those in an untagged nucleoside. We now introduce the use of an indirect tag at the C8 of 15N7-labeled purine nucleosides. This tag allows unambiguous differentiation between a pair of 15N7-labeled purines in which only one is 13C8 labeled. Although the very small C8-N7 coupling (<1 Hz) precludes its direct detection in 1D 15N spectra, 2D 1H-15N NMR experiments display the large C8-H8 coupling (>200 Hz) because H8 is coupled to both N7 and C8. The 13C8 atom is introduced by means of a ring closure of the exocyclic amino groups of a pyrimidinone using [13C]sodium ethyl xanthate. Here, we present methods for the syntheses of [8-13C-1,7,NH2-15N3]adenosine, -guanosine, and their deoxy analogues.
Sigma-Aldrich
Potassium ethyl xanthogenate, 96%