Merck
Syntheses of specifically deoxygenated methyl alpha-isomaltotriosides.
Carbohydrate research (1992-09-02)
E Petráková, P Kovác, C P Glaudemans
ABSTRACT
Specifically deoxygenated methyl alpha-isomaltotriosides were synthesized by the silver perchlorate-mediated condensation of a protected alpha-isomaltosyl chloride with suitably blocked derivatives of methyl alpha-D-glucopyranoside deoxygenated respectively at positions 2, 3, and 4, followed by deprotection.
Sigma-Aldrich
Silver perchlorate, anhydrous, 97%