Merck
  • Rapid analysis of terpene lactones in extract of Ginkgo biloba L. by high performance liquid chromatography.
Rapid analysis of terpene lactones in extract of Ginkgo biloba L. by high performance liquid chromatography.
Se pu = Chinese journal of chromatography (2003-01-25)
H F Wang, X R Ju
ABSTRACT
A new rapid analytical method was developed for the quantification of terpene lactones (bilobalide(BB) and ginkgolide A, B, C, J) in extract of Ginkgo biloba L. (EGb) using a liquid-liquid solvent extraction procedure followed by high performance liquid chromatography. EGb was dissolved in 30% ethanol and extracted with ether. After evaporation, the residue was then determined by HPLC on a C18 column with methanol-water-orthophosphoric acid (25:75:0.1, V/V) as eluent and refractive index (RI) detection. Results showed that the excellent sample clean-up procedure is more simple and specific, and saves more time (less than 20 min) than any other methods that have been reported, and also leads to high recoveries (> 99.0%) and low RSDs (< 2.0%). The reproducible method is regarded to be very useful for evaluating the quality of extract of Ginkgo biloba L.