Merck
Removal of selenocyanate from water using elemental iron.
Water research (2002-10-09)
Xiaoguang Meng, Sunbaek Bang, George P Korfiatis
ABSTRACT
Batch experiments were conducted to investigate the removal of selenocyanate (SeCN-) from oil refinery wastewater and artificial wastewater with elemental iron [Fe(0)]. The chemical forms of selenium in the reacted solids were determined with X-ray photoelectron spectroscopy (XPS) and a sulfite extraction procedure. SeCN- was effectively removed from the wastewater with Fe(0) filings when the water pH was controlled at approximately 6. SeCN- was removed by Fe(0) through the formation of elemental selenium [Se(0)] and ferrous selenide. The possible chemical reactions between SeCN- and Fe(0) included deselenation of SeCN- and electrochemical reduction of Se(0) to selenide. A cost-effective process may be developed for the treatment of SeCN- in wastewater using Fe(0).
Sigma-Aldrich
Potassium selenocyanate, reagent grade, 97%
Sigma-Aldrich
Potassium selenocyanate, ReagentPlus®, ≥99%