Merck
  • Laboratory studies to assess the risk of development of resistance to zoxamide.
Laboratory studies to assess the risk of development of resistance to zoxamide.
Pest management science (2001-11-28)
D H Young, S L Spiewak, R A Slawecki
ABSTRACT
Laboratory studies were conducted to evaluate the risk of developing field resistance to zoxamide, a new Oomycete fungicide which acts on microtubules. Zoxamide, metalaxyl and dimethomorph were compared with respect to the ease with which fungicide-resistant mutants could be isolated and their level of resistance. Attempts to generate mutants of Phytophthora capsici and P infestans with resistance to zoxamide by mycelial adaptation on fungicide-amended medium were unsuccessful. Similarly, changes in sensitivity to zoxamide were small (resistance factors < or = 2.2) in mutants of P capsici isolated by chemical mutagenesis of zoospore cysts. In parallel experiments with metalaxyl, highly resistant mutants were obtained using both adaptation (P capsici or P infestans) and chemical mutagenesis (P capsici). For dimethomorph, chemical mutagenesis (P capsici) yielded moderately resistant mutants (maximum resistance factor = 20.9), and adaptation (P capsici or P infestans) did not induce resistance. It is proposed that failure to isolate mutants resistant to zoxamide results from the diploid nature of Oomycete fungi and the likelihood that target-site mutations would produce a recessive phenotype. Our studies suggest that the risk of a highly resistant pathogen population developing rapidly in the field is much lower for zoxamide than for metalaxyl. However, as with any site-specific fungicide, appropriate precautions against resistance development should be taken.
Product Description
Supelco
Dimethomorph, PESTANAL®, analytical standard