Merck
  • Synthesis of styrenes through the biocatalytic decarboxylation of trans-cinnamic acids by plant cell cultures.
Synthesis of styrenes through the biocatalytic decarboxylation of trans-cinnamic acids by plant cell cultures.
Chemical & pharmaceutical bulletin (2001-06-01)
M Takemoto, K Achiwa
ABSTRACT
A novel method for producing styrenes from trans-cinnamic acids was developed. When trans-cinnamic acid was incubated with plant cell cultures at room temperature, styrene was obtained. 4-Hydroxy-3-methoxystyrene (2a), 3-nitrostyrene (2f) and furan (2g) were synthesized quantitatively.
Sigma-Aldrich
2-Methoxy-4-vinylphenol, ≥98%, FG
Sigma-Aldrich
trans-Cinnamic acid, ≥99%