Merck
  • Geranic acid formation, an initial reaction of anaerobic monoterpene metabolism in denitrifying Alcaligenes defragrans.
Geranic acid formation, an initial reaction of anaerobic monoterpene metabolism in denitrifying Alcaligenes defragrans.
Applied and environmental microbiology (2000-07-06)
U Heyen, J Harder
ABSTRACT
Monoterpenes with an unsaturated hydrocarbon structure are mineralized anaerobically by the denitrifying beta-proteobacterium Alcaligenes defragrans. Organic acids occurring in cells of A. defragrans and culture medium were characterized to identify potential products of the monoterpene activation reaction. Geranic acid (E,E-3,7-dimethyl-2,6-octadienoic acid) accumulated to 0.5 mM in cells grown on alpha-phellandrene under nitrate limitation. Cell suspensions of A. defragrans 65Phen synthesized geranic acid in the presence of beta-myrcene, alpha-phellandrene, limonene, or alpha-pinene. Myrcene yielded the highest transformation rates. The alicyclic acid was consumed by cell suspensions during carbon limitation. Heat-labile substances present in cytosolic extracts catalyzed the formation of geranic acid from myrcene. These results indicated that a novel monoterpene degradation pathway must be present in A. defragrans.
Sigma-Aldrich
Geranic acid, technical grade, 85%
Sigma-Aldrich
Geranic acid, 85%, stabilised