Merck
  • Neurons projecting to the retrocerebral complex of the adult blow fly, Protophormia terraenovae.
Neurons projecting to the retrocerebral complex of the adult blow fly, Protophormia terraenovae.
Cell and tissue research (2000-04-20)
S Shiga, I Toyoda, H Numata
ABSTRACT
Anatomical study of neurons projecting to the retrocerebral complex of the adult blow fly, Protophormia terraenovae, was done by NiCl2 filling and immunocytochemistry. Retrograde filling through the cardiac-recurrent nerve labeled three groups of neurons in the brain/subesophageal ganglion: (1) paramedial clusters of the pars intercerebralis, (2) neurons in each pars lateralis, and (3) neurons in the subesophageal ganglion. The pars intercerebralis neurons send prominent axons into the median bundle and exit from the brain via the contralateral nervus corporis cardiaci. Based on the projection pattern, two types of the pars lateralis neurons can be distinguished: the most lateral pairs of neurons contralaterally extend through the posterior lateral tract and the remainder ipsilaterally extend through the posterior lateral tract. The neurons in the subesophageal ganglion run through the contralateral nervus corporis cardiaci. The dendritic arborization of the pars intercerebralis and pars lateralis neurons is restricted to the superior protocerebral neuropil and to the anterior neuropil of the subesophageal ganglion where the neurons in the subesophageal ganglion also project. Retrograde filling from the corpus allatum indicated that the pars lateralis neurons and a few pars intercerebralis neurons project to the corpus allatum, but that the neurons in the subesophageal ganglion do not. Orthograde filling from the pars intercerebralis and staining by paraldehyde-thionin/paraldehyde-fuchsin indicated that the pars intercerebralis neurons project primarily to the corpus cardiacum/hypocerebral ganglion complex. Immunostaining with a polyclonal antiserum against diapause hormone, a member of the FXPRLamide family, suggests that some of the subesophageal ganglion neurons contain FXPRLamide-like peptides.
Sigma-Aldrich
Paraldehyde, ≥97%
Sigma-Aldrich
Paraldehyde, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Paraldehyde, ≥97.0% (GC)