Merck
  • Signal transducer and activator of transcription 3 (STAT3) regulates collagen-induced platelet aggregation independently of its transcription factor activity.
Signal transducer and activator of transcription 3 (STAT3) regulates collagen-induced platelet aggregation independently of its transcription factor activity.
Circulation (2012-12-26)
Zhou Zhou, Francisca C Gushiken, Doug Bolgiano, Breia J Salsbery, Niloufar Aghakasiri, Naijie Jing, Xiaoping Wu, K Vinod Vijayan, Rolando E Rumbaut, Roberto Adachi, Jose A Lopez, Jing-Fei Dong
ABSTRACT
Platelet hyperactivity induced by inflammation is a known risk factor for atherosclerosis and thrombosis, but its underlying mechanisms remain poorly understood. The signal transducer and activator of transcription 3 (STAT3) was activated in collagen-stimulated platelets. Activated STAT3 served as a protein scaffold to facilitate the catalytic interaction between the kinase Syk (spleen tyrosine kinase) and the substrate PLCγ2 to enhance collagen-induced calcium mobilization and platelet activation. The same interaction of STAT3 with Syk and PLCγ2 was detected in HEK293 cells transfected with cDNAs for Syk and PLCγ2 and stimulated with interleukin-6. Pharmacological inhibition of STAT3 blocked ≈50% of collagen- and a collagen-related peptide-induced but not thrombin receptor-activating peptide- or ADP-induced aggregation and ≈80% of thrombus formation of human platelets on a collagen matrix. This in vitro phenotype was reproduced in mice infused with STAT3 inhibitors and mice with platelet-specific STAT3 deficiency. By forming a complex with its soluble receptor, the proinflammatory cytokine interleukin-6 enhanced the collagen-induced STAT3 activation in human platelets. These data demonstrate a nontranscriptional activity of STAT3 that facilitates a crosstalk between proinflammatory cytokine and hemostasis/thrombosis signals in platelets. This crosstalk may be responsible for the platelet hyperactivity found in conditions of inflammation.
Sigma-Aldrich
Phospholipase C from Clostridium perfringens (C. welchii), Type XIV, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
Phospholipase C from Clostridium perfringens (C. welchii), Type I, lyophilized powder, 10-50 units/mg protein
Sigma-Aldrich
Phospholipase C from Bacillus cereus, ≥200 units/mg protein