Merck
  • Effects of pharmaceutical processing on pepsin activity during the formulation of solid dosage forms.
Effects of pharmaceutical processing on pepsin activity during the formulation of solid dosage forms.
Pharmaceutical development and technology (2012-12-12)
Katalin Kristó, Klára Pintye-Hódi
ABSTRACT
The main aim of this study was to investigate the effects of pharmaceutical technological methods on pepsin activity during the formulation of solid dosage forms. The circumstances of direct compression and wet granulation were modeled. During direct compression, the heat and the compression force must be taken into consideration. The effects of these parameters were investigated in three materials (pure pepsin, and 1:1 (w/w) pepsin-tartaric acid and 1:1 (w/w) pepsin-citric acid powder mixtures). It was concluded that direct compression is appropriate for the formulation of solid dosage forms containing pepsin through application without acids or with acids at low compression force. The effects of wet granulation were investigated with a factorial design for the same three materials. The factors were time, temperature and moisture content. There was no significant effect of the factors when acids were not applied. Temperature was a significant factor when acids were applied. The negative effect was significantly higher for citric acid than for tartaric acid. It was found that wet granulation can be utilized for the processing of pepsin into solid dosage forms under well-controlled circumstances. The application of citric acid is not recommended during the formulation of solid dosage forms through wet granulation. A mathematically based optimization may be necessary for preformulation studies of the preparation of dosage forms containing sensitive enzymes.
Sigma-Aldrich
DL-Tartaric acid, ≥99%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
DL-Tartaric acid, ReagentPlus®, 99%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Supelco
L-(+)-Tartaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-(−)-Tartaric acid, ReagentPlus®, 99%
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.7%, FCC, FG
Sigma-Aldrich
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
L-(+)-Tartaric acid, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
D-(−)-Tartaric acid, puriss., unnatural form, ≥99.0% (T)
Sigma-Aldrich
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, BioXtra
Sigma-Aldrich
L-(+)-Tartaric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%