Merck
  • Aberrant promoter methylation profile of Niemann-pick type C1 gene in cardiovascular disease.
Aberrant promoter methylation profile of Niemann-pick type C1 gene in cardiovascular disease.
Iranian biomedical journal (2013-04-10)
Masoumeh Afzali, Alireza Nakhaee, Seyed Payman Tabatabaei, Kourosh Tirgar-Fakheri, Mohammad Hashemi
ABSTRACT
The protein of Niemann-pick type C1 (NPC1) gene promotes the egress of cholesterol from late endosomes and lysosomes to other cellular compartments and contributes to a process known as reverse cholesterol transport. This study aimed to examine whether promoter methylation of NPC1 is associated with risk of cardiovascular disease (CVD). Fifty CVD patients and 50 healthy subjects as the control group were recruited in this study. Promoter methylation of NPC1 gene was defined using a nested-methylation specific polymerase chain reaction method. Statistical analyses were done using the chi-square, t-test or ANOVA tests. Our study showed that the frequency of semi-methylated promoter (methylated/unmethylated status) was significantly higher in CVD patients than that in controls (OR = 6.521, 95% CI = 2.211-19.215, P = 0.008). However, a completely methylated promoter (methylated/methylated status) was not detected in any subjects in either of the two groups tested. Additionally, the analysis of clinical data according to the methylation status of NPC1 gene demonstrated that serum levels of total cholesterol, total triglycerides, high low-density lipoprotein cholesterol (LDL-C) and low high-density lipoprotein cholesterol (HDL-C) are influenced by NPC1 methylation, so that subjects with a completely unmethylated promoter (Unmethylated/unmethylated status) held lower levels of total triglycerides, total cholesterol, LDL-C and higher levels of HDL-C. Our findings propose that the NPC1 promoter methylation is a probable mechanism that can result in reduced/impaired NPC1 expression/activity and may thus contribute to progression of CVD.
Sigma-Aldrich
Imprint® DNA Modification Kit, For bisulfite DNA conversion & purification