Merck
  • Acute effects of decaffeinated coffee and the major coffee components chlorogenic acid and trigonelline on glucose tolerance.
Acute effects of decaffeinated coffee and the major coffee components chlorogenic acid and trigonelline on glucose tolerance.
Diabetes care (2009-03-28)
Aimée E van Dijk, Margreet R Olthof, Joke C Meeuse, Elin Seebus, Rob J Heine, Rob M van Dam
ABSTRACT
Coffee consumption has been associated with lower risk of type 2 diabetes. We evaluated the acute effects of decaffeinated coffee and the major coffee components chlorogenic acid and trigonelline on glucose tolerance. We conducted a randomized crossover trial of the effects of 12 g decaffeinated coffee, 1 g chlorogenic acid, 500 mg trigonelline, and placebo (1 g mannitol) on glucose and insulin concentrations during a 2-h oral glucose tolerance test (OGTT) in 15 overweight men. Chlorogenic acid and trigonelline ingestion significantly reduced glucose (-0.7 mmol/l, P = 0.007, and -0.5 mmol/l, P = 0.024, respectively) and insulin (-73 pmol/l, P = 0.038, and -117 pmol/l, P = 0.007) concentrations 15 min following an OGTT compared with placebo. None of the treatments affected insulin or glucose area under the curve values during the OGTT compared with placebo. Chlorogenic acid and trigonelline reduced early glucose and insulin responses during an OGTT.
Product Description
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)