Merck
  • Ginsenoside Rg1 attenuates lipopolysaccharide-induced chronic liver damage by activating Nrf2 signaling and inhibiting inflammasomes in hepatic cells.
Ginsenoside Rg1 attenuates lipopolysaccharide-induced chronic liver damage by activating Nrf2 signaling and inhibiting inflammasomes in hepatic cells.
Journal of ethnopharmacology (2024-01-21)
Huimin Zhou, Yan Liu, Yong Su, Pengmin Ji, Liangliang Kong, Ran Sun, Duoduo Zhang, Hanyang Xu, Weiping Li, Weizu Li
ABSTRACT
Ginseng (Panax ginseng C. A. Meyer) is a precious traditional Chinese medicine with multiple pharmacological effects. Ginsenoside Rg1 is a main active ingredient extracted from ginseng, which is known for its age-delaying and antioxidant effects. Increasing evidence indicates that Rg1 exhibits anti-inflammatory properties in numerous diseases and may ameliorate oxidative damage and inflammation in many chronic liver diseases. Chronic inflammatory injury in liver cells is an important pathological basis of many liver diseases. However, its mechanism remains unclear and therapeutic strategies to prevent its development need to be further explored. Thus, our study is to delve the protective effect and mechanism of Rg1 against chronic hepatic inflammatory injuries induced by lipopolysaccharide (LPS). The chronic liver damage model in mice was build up by injecting intraperitoneally with LPS (200 μg/kg) for 21 days. Serum liver function indicators and levels of IL-1β, IL-6 and TNF-α were examined by using corresponding Kits. Hematoxylin and Eosin (H&E), Periodic acid-Schiff (PAS), and Masson stains were utilized to visualize hepatic histopathological damage, glycogen deposition, and liver fibrosis. The nuclear import of p-Nrf2 and the generation of Col4 in the liver were detected by IF, while IHC was employed to detect the expressions of NLRP3 and AIM2 in the hepatic. The Western blot and q-PCR were used to survey the expressions of proteins and mRNAs of fibrosis and apoptosis, and the expressions of Keap1, p-Nrf2 and NLRP3, NLRP1, AIM2 inflammasome-related proteins in mouse liver. The cell viability of human hepatocellular carcinoma cells (HepG2) was detected by Cell Counting Kit-8 to select the action concentration of LPS, and intracellular ROS generation was detected using a kit. The expressions of Nuclear Nrf2, HO-1, NQO1 and NLRP3, NLRP1, and AIM2 inflammasome-related proteins in HepG2 cells were detected by Western blot. Finally, the feasibility of the molecular interlinking between Rg1 and Nrf2 was demonstrated by molecular docking. Rg1 treatment for 21 days decreased the levels of ALT, AST, and inflammatory factors of serum IL-1β, IL-6 and TNF-α in mice induced by LPS. Pathological results indicated that Rg1 treatment obviously alleviated hepatocellular injury and apoptosis, inflammatory cell infiltration and liver fibrosis in LPS stimulated mice. Rg1 promoted Keap1 degradation and enhanced the expressions of p-Nrf2, HO-1 and decreased the levels of NLRP1, NLRP3, AIM2, cleaved caspase-1, IL-1β and IL-6 in livers caused by LPS. Furthermore, Rg1 effectively suppressed the rise of ROS in HepG2 cells induced by LPS, whereas inhibition of Nrf2 reversed the role of Rg1 in reducing the production of ROS and NLRP3, NLRP1, and AIM2 expressions in LPS-stimulated HepG2 cells. Finally, the molecular docking illustrated that Rg1 exhibits a strong affinity towards Nrf2. The findings indicate that Rg1 significantly ameliorates chronic liver damage and fibrosis induced by LPS. The mechanism may be mediated through promoting the dissociation of Nrf2 from Keap1 and then activating Nrf2 signaling and further inhibiting NLRP3, NLRP1, and AIM2 inflammasomes in liver cells.
Sigma-Aldrich
Ginsenoside Rg1, ≥98% (HPLC)