Merck
  • Ferrostatin-1 attenuates brain injury in animal model of subarachnoid hemorrhage via phospholipase A2 activity of PRDX6.
Ferrostatin-1 attenuates brain injury in animal model of subarachnoid hemorrhage via phospholipase A2 activity of PRDX6.
Neuroreport (2023-07-03)
Huiqing Wang, Yao Zhou, Mingpei Zhao, Lianghong Yu, Yuanxiang Lin, Dezhi Kang
ABSTRACT
Subarachnoid hemorrhage (SAH) is an acute catastrophic neurological disorder with high morbidity and mortality. Ferroptosis is one of the pathophysiological processes during secondary brain injury of SAH, which could be inhibited by ferrostatin-1 (Fer-1) effectively. Peroxiredoxin6 (PRDX6) is an antioxidant protein and is currently proven to be associated with lipid peroxidation in ferroptosis except in GSH/GPX4 and FSP1/CoQ10 antioxidant systems. However, the alteration and function of PRDX6 in SAH are still unknown. In addition, whether PRDX6 is involved in the neuroprotection of Fer-1 in SAH is yet to be investigated. Endovascular perforation was employed to induce the SAH model. Fer-1 and in vivo siRNA aiming to knockdown PRDX6 were administrated intracerebroventricularly to investigate relevant regulation and mechanism. We confirmed the inhibition of ferroptosis and neuroprotection from brain injury by Fer-1 in SAH. The induction of SAH reduced the expression of PRDX6, which could be alleviated by Fer-1. Accordingly, dysregulated lipid peroxidation indicated by GSH and MDA was improved by Fer-1, which was counteracted by si-PRDX6. Similarly, the neuroprotection of Fer-1 in SAH was diminished by the knockdown of PRDX6 and the administration of a calcium-independent phospholipase A2 (iPLA2) inhibitor. PRDX6 is involved in ferroptosis induced by SAH and is associated with Fer-1 neuroprotection from brain injury via its iPLA2 activity.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-PRDX6 (AB1) antibody produced in rabbit, affinity isolated antibody