MilliporeSigma
  • Targeting of voltage-gated calcium channel α2δ-1 subunit to lipid rafts is independent from a GPI-anchoring motif.
Targeting of voltage-gated calcium channel α2δ-1 subunit to lipid rafts is independent from a GPI-anchoring motif.
PloS one (2011-06-23)
Philip Robinson, Sarah Etheridge, Lele Song, Riddhi Shah, Elizabeth M Fitzgerald, Owen T Jones
ABSTRACT
Voltage-gated calcium channels (Ca(v)) exist as heteromultimers comprising a pore-forming α(1) with accessory β and α(2)δ subunits which modify channel trafficking and function. We previously showed that α(2)δ-1 (and likely the other mammalian α(2)δ isoforms--α(2)δ-2, 3 and 4) is required for targeting Ca(v)s to lipid rafts, although the mechanism remains unclear. Whilst originally understood to have a classical type I transmembrane (TM) topology, recent evidence suggests the α(2)δ subunit contains a glycosylphosphatidylinositol (GPI)-anchor that mediates its association with lipid rafts. To test this notion, we have used a strategy based on the expression of chimera, where the reported GPI-anchoring sequences in the gabapentinoid-sensitive α(2)δ-1 subunit have been substituted with those of a functionally inert Type I TM-spanning protein--PIN-G. Using imaging, electrophysiology and biochemistry, we find that lipid raft association of PIN-α(2)δ is unaffected by substitution of the GPI motif with the TM domain of PIN-G. Moreover, the presence of the GPI motif alone is not sufficient for raft localisation, suggesting that upstream residues are required. GPI-anchoring is susceptible to phosphatidylinositol-phospholipase C (PI-PLC) cleavage. However, whilst raft localisation of PIN-α(2)δ is disrupted by PI-PLC treatment, this is assay-dependent and non-specific effects of PI-PLC are observed on the distribution of the endogenous raft marker, caveolin, but not flotillin. Taken together, these data are most consistent with a model where α(2)δ-1 retains its type I transmembrane topology and its targeting to lipid rafts is governed by sequences upstream of the putative GPI anchor, that promote protein-protein, rather than lipid-lipid interactions.
Sigma-Aldrich
Cell Dissociation Solution Non-enzymatic 1x, Prepared in phosphate buffered saline without calcium and magnesium, sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cell Dissociation Solution Non-enzymatic 1x, Prepared in Hanks′ Balanced Salt Solution without calcium and magnesium, sterile-filtered, BioReagent, suitable for cell culture