Merck
  • Binucleated human hepatocytes arise through late cytokinetic regression during endomitosis M phase.
Binucleated human hepatocytes arise through late cytokinetic regression during endomitosis M phase.
The Journal of cell biology (2024-05-10)
Gabriella S Darmasaputra, Cindy C Geerlings, Susana M Chuva de Sousa Lopes, Hans Clevers, Matilde Galli
ABSTRACT
Binucleated polyploid cells are common in many animal tissues, where they arise by endomitosis, a non-canonical cell cycle in which cells enter M phase but do not undergo cytokinesis. Different steps of cytokinesis have been shown to be inhibited during endomitosis M phase in rodents, but it is currently unknown how human cells undergo endomitosis. In this study, we use fetal-derived human hepatocyte organoids (Hep-Orgs) to investigate how human hepatocytes initiate and execute endomitosis. We find that cells in endomitosis M phase have normal mitotic timings, but lose membrane anchorage to the midbody during cytokinesis, which is associated with the loss of four cortical anchoring proteins, RacGAP1, Anillin, SEPT9, and citron kinase (CIT-K). Moreover, reduction of WNT activity increases the percentage of binucleated cells in Hep-Orgs, an effect that is dependent on the atypical E2F proteins, E2F7 and E2F8. Together, we have elucidated how hepatocytes undergo endomitosis in human Hep-Orgs, providing new insights into the mechanisms of endomitosis in mammals.
Sigma-Aldrich
Anti-Anillin Antibody, clone 5f3.1, clone 5F3.1, from mouse
Sigma-Aldrich
Anti-SEPT9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution