Merck
  • C9orf72 proline-arginine dipeptide repeats disrupt the proteasome and perturb proteolytic activities.
C9orf72 proline-arginine dipeptide repeats disrupt the proteasome and perturb proteolytic activities.
Journal of neuropathology and experimental neurology (2023-10-04)
Yifan Zhang, Sophia C K Nelson, Ashley P Viera Ortiz, Edward B Lee, Robert Fairman
ABSTRACT
The hexanucleotide G4C2 repeat expansion in C9orf72 is the most frequent genetic cause of familial amyotrophic lateral sclerosis (ALS). Aberrant translation of this hexanucleotide sequence leads to production of 5 dipeptide repeats (DPRs). One of these DPRs is proline-arginine (polyPR), which is found in C9orf72-expanded ALS (C9ALS) patient brain tissue and is neurotoxic across multiple model systems. PolyPR was previously reported to bind and impair proteasomes in vitro. Nevertheless, the clinical relevance of the polyPR-proteasome interaction and its functional consequences in vivo are yet to be established. Here, we aim to confirm and functionally characterize polyPR-induced impairment of proteolysis in C9ALS patient tissue and an in vivo model system. Confocal microscopy and immunofluorescence studies on both human and Drosophila melanogaster brain tissues revealed sequestration of proteasomes by polyPR into inclusion-like bodies. Co-immunoprecipitation in D. melanogaster showed that polyPR strongly binds to the proteasome. In vivo, functional evidence for proteasome impairment is further shown by the accumulation of ubiquitinated proteins along with lysosomal accumulation and hyper-acidification, which can be rescued by a small-molecule proteasomal enhancer. Together, we provide the first clinical report of polyPR-proteasome interactions and offer in vivo evidence proposing polyPR-induced proteolytic dysfunction as a pathogenic mechanism in C9ALS.
Sigma-Aldrich
Anti-C9ORF72/C9RANT (Poly-PR), serum, from rabbit