Merck
  • Evaluating smartphone-based optical readouts for immunoassays in human and veterinary healthcare: A comparative study.
Evaluating smartphone-based optical readouts for immunoassays in human and veterinary healthcare: A comparative study.
Talanta (2024-04-23)
Melania Mesas Gómez, Esther Julián, Lara Armengou, Maria Isabel Pividori
ABSTRACT
Recent advances have significantly enhanced the use of smartphone devices for medical diagnostics. This study uses high-resolution cameras in mobile devices to capture and process bioassay images, enabling the quantification of diverse biomarkers across a range of diagnostic tests conducted on 96-well microplates. The study evaluates the effectiveness of this technology through protein quantification techniques and immunoassays that generate colorimetric responses at specific wavelengths. It includes the assessment of bicinchoninic acid and Bradford protein quantification methods, alongside a conventional immunoassay for detecting mare antibodies in colostrum to monitor foal immunodeficiencies. Further application involves the readout of magneto-actuated immunoassays aimed at quantifying bacteria. The results obtained from benchtop spectrophotometry at 595, 562, and 450 nm are compared with those acquired using a smartphone, which identified color intensities in shades of blue, purple, and yellow. This comparison yields promising correlations for the samples tested, suggesting a high degree of accuracy in the smartphone capability to analyze bioassay outcomes. The analysis via smartphone is facilitated by a specific app, which processes the images captured by the phone camera to quantify color intensities corresponding to different biomarker concentrations. Detection limits of 12.3 and 22.8 μg mL-1 for the bicinchoninic acid assay and 36.7 and 45.4 μg mL-1 for the Bradford are obtained for protein quantification using the spectrophotometer and the smartphone app, respectively. For mare's antibodies in colostrum, the values are 1.14 and 1.72 ng mL-1, while the detection of E. coli is performed at 2.0 x 104 and 2.9 × 104 CFU mL-1, respectively. This approach offers further advantages, including wide availability, cost-effectiveness, portability, compared to traditional and expensive benchtop instruments.
Sigma-Aldrich
Anti-Horse IgG (Fc specific) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.