Merck
  • Innate immune cell activation after HIV-1 vaccine administration is associated with increased antibody production.
Innate immune cell activation after HIV-1 vaccine administration is associated with increased antibody production.
Frontiers in immunology (2024-02-29)
Kombo F N'guessan, Kawthar Machmach, Isabella Swafford, Margaret C Costanzo, Lindsay Wieczorek, Dohoon Kim, Siriwat Akapirat, Victoria R Polonis, Punnee Pitisuttithum, Sorachai Nitayaphan, Sanjay Gurunathan, Faruk Sinangil, Suwat Chariyalertsak, Julie A Ake, Robert J O'connell, Sandhya Vasan, Dominic Paquin-Proulx
ABSTRACT
The RV144 Thai phase III clinical trial's canarypox-protein HIV vaccine regimen showed modest efficacy in reducing infection. We therefore sought to determine the effects of vaccine administration on innate cell activation and subsequent associations with vaccine-induced immune responses. RV306 was a randomized, double-blind clinical trial in HIV-uninfected Thai adults that tested delayed boosting following the RV144 regimen. PBMC collected from RV306 participants prior to and 3 days after the last boost were used to investigate innate immune cell activation. Our analysis showed an increase in CD38+ mucosal associated invariant T (MAIT) cells, CD38+ invariant natural killer T (iNKT) cells, CD38+ γδ T cells, CD38+, CD69+ and HLA-DR+ NK cells 3 days after vaccine administration. An increase in CD14-CD16+ non-classical monocytes and CD14+CD16+ intermediate monocytes accompanied by a decrease in CD14+CD16- classical monocytes was also associated with vaccine administration. Inclusion of ALVAC-HIV in the boost did not further increase MAIT, iNKT, γδ T, and NK cell activation or increase the proportion of non-classical monocytes. Additionally, NK cell activation 3 days after vaccination was positively associated with antibody titers of HIV Env-specific total IgG and IgG1. Vδ1 T cell activation 3 days after vaccine administration was associated with HIV Env-specific IgG3 titers. Finally, we observed trending associations between MAIT cell activation and Env-specific IgG3 titers and between NK cell activation and TH023 pseudovirus neutralization titers. Our study identifies a potential role for innate cells, specifically NK, MAIT, and γδ T cells, in promoting antibody responses following HIV-1 vaccine administration.
Sigma-Aldrich
Milli-Mark® Anti-FcεRI Antibody, γ subunit-FITC, Milli-Mark®, from rabbit
Pricing and availability is not currently available.