Merck
  • Multidose pharmacokinetics of orally administered florfenicol in the channel catfish (Ictalurus punctatus).
Multidose pharmacokinetics of orally administered florfenicol in the channel catfish (Ictalurus punctatus).
Journal of veterinary pharmacology and therapeutics (2012-08-14)
P S Gaunt, C Langston, C Wrzesinski, D Gao, P Adams, L Crouch, D Sweeney, R Endris
ABSTRACT
Plasma disposition of florfenicol in channel catfish was investigated after an oral multidose (10 mg/kg for 10 days) administration in freshwater at water temperatures ranging from 24.7 to 25.9 °C. Florfenicol concentrations in plasma were analyzed by means of liquid chromatography with MS/MS detection. After the administration of florfenicol, the mean terminal half-life (t(1/2)), maximum concentration at steady-state (Css (max)), time of Css (max) (T(max)), minimal concentration at steady-state (Css (min)), and Vc /F were 9.0 h, 9.72 μg/mL, 8 h, 2.53 μg/mL, and 0.653 L/kg, respectively. These results suggest that florfenicol administered orally at 10 mg/kg body weight for 10 days could be expected to control catfish bacterial pathogens inhibited in vitro by a minimal inhibitory concentration value of <2.5 μg/mL.
Sigma-Aldrich
Florfenicol, ≥98% (HPLC)