Merck
  • Daxx knockdown promoted rDNA transcription without impairing H3.3 expression in mouse preimplantation embryos.
Daxx knockdown promoted rDNA transcription without impairing H3.3 expression in mouse preimplantation embryos.
Anatomia, histologia, embryologia (2023-09-28)
Sihang Song, Nan Wang, Xing-Wei Huang, Qi Jiang, Xiang-Rong Cheng, Nan Pang, Lei Lei
ABSTRACT
During fertilization, DAXX (death domain-associated protein) mediates histone variant H3.3 incorporation into heterochromatin, which plays an important role in the maintenance of genomic integrity. rDNA, the ribosomal gene, is included in the first wave of gene activation after fertilization. Our and other studies indicated that loss of Daxx disturbs rDNA heterochromatinization and promotes rDNA transcription without change in protein expression of H3.3. However, maternal and zygotic deletion of Daxx impairs blastocyst development. Whether Daxx knockdown affects H3.3 expression and improves the rDNA transcription in preimplantation development has not been reported. In the present study, we injected HA-labelled H3.3 (H3.3-HA) into oocytes during ICSI procedure, and detected H3.3 and DAXX by immunofluorescent staining. Then, we knockdowned Daxx and detected the gene expression levels of Daxx, H3.3, 18s and 47s rRNA. We also performed immunofluorescent staining of B23, γH2A and EdU incorporation to demonstrate nuclear structure, DNA damage and replication. We found injection of H3.3-HA did not impair preimplantation development. Daxx siRNA did not change expression of H3.3 mRNA, and the development of two-cell embryos and blastocysts, but the overall replication and expression levels of rRNA were increased compared with that in the control group. Finally, knockdown of DAXX did not aggravate the DNA damage but loosened the nucleolus. We concluded that Daxx knockdown promoted DNA replication and rDNA transcription, but did not affect H3.3 expression and subsequent preimplantation development.
Sigma-Aldrich
Rabbit anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Anti-DAXX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution