Merck
  • Beta-2 adrenergic receptor agonism alters astrocyte phagocytic activity and has potential applications to psychiatric disease.
Beta-2 adrenergic receptor agonism alters astrocyte phagocytic activity and has potential applications to psychiatric disease.
Discover mental health (2023-12-01)
Ellen R Bowen, Phillip DiGiacomo, Hannah P Fraser, Kevin Guttenplan, Benjamin A H Smith, Marlene L Heberling, Laura Vidano, Nigam Shah, Mehrdad Shamloo, Jennifer L Wilson, Kevin V Grimes
ABSTRACT
Schizophrenia is a debilitating condition necessitating more efficacious therapies. Previous studies suggested that schizophrenia development is associated with aberrant synaptic pruning by glial cells. We pursued an interdisciplinary approach to understand whether therapeutic reduction in glial cell-specifically astrocytic-phagocytosis might benefit neuropsychiatric patients. We discovered that beta-2 adrenergic receptor (ADRB2) agonists reduced phagocytosis using a high-throughput, phenotypic screen of over 3200 compounds in primary human fetal astrocytes. We used protein interaction pathways analysis to associate ADRB2, to schizophrenia and endocytosis. We demonstrated that patients with a pediatric exposure to salmeterol, an ADRB2 agonist, had reduced in-patient psychiatry visits using a novel observational study in the electronic health record. We used a mouse model of inflammatory neurodegenerative disease and measured changes in proteins associated with endocytosis and vesicle-mediated transport after ADRB2 agonism. These results provide substantial rationale for clinical consideration of ADRB2 agonists as possible therapies for patients with schizophrenia.
Sigma-Aldrich
Anti-phospho-DAP-Kinase (pSer308) antibody, Mouse monoclonal, clone DKPS308, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Anti-phospho-Synapsin I (Ser62,Ser67) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone SP15 (Ascites Free), clone SP15, from mouse