Merck
  • Excessive negative regulation of type I interferon disrupts viral control in individuals with Down syndrome.
Excessive negative regulation of type I interferon disrupts viral control in individuals with Down syndrome.
Immunity (2022-10-16)
Louise Malle, Marta Martin-Fernandez, Sofija Buta, Ashley Richardson, Douglas Bush, Dusan Bogunovic
ABSTRACT
Down syndrome (DS) is typically caused by triplication of chromosome 21. Phenotypically, DS presents with developmental, neurocognitive, and immune features. Epidemiologically, individuals with DS have less frequent viral infection, but when present, these infections lead to more severe disease. The potent antiviral cytokine type I Interferon (IFN-I) receptor subunits IFNAR1 and IFNAR2 are located on chromosome 21. While increased IFNAR1/2 expression initially caused hypersensitivity to IFN-I, it triggered excessive negative feedback. This led to a hypo-response to subsequent IFN-I stimuli and an ensuing viral susceptibility in DS compared to control cells. Upregulation of IFNAR2 expression phenocopied the DS IFN-I dynamics independent of trisomy 21. CD14+ monocytes from individuals with DS exhibited markers of prior IFN-I exposure and had muted responsiveness to ex vivo IFN-I stimulation. Our findings unveil oscillations of hyper- and hypo-response to IFN-I in DS, predisposing individuals to both lower incidence of viral disease and increased infection-related morbidity and mortality.
Sigma-Aldrich
Anti-IFNAR-1 Antibody, clone MARI-5A3, clone MARI-5A3, from mouse
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-STAT2 Antibody, CT, Upstate®, from rabbit