Merck
  • A family of carboxypeptidases catalyzing α- and β-tubulin tail processing and deglutamylation.
A family of carboxypeptidases catalyzing α- and β-tubulin tail processing and deglutamylation.
Science advances (2023-09-13)
Simon Nicot, Ghislain Gillard, Hathaichanok Impheng, Ewa Joachimiak, Serge Urbach, Kazufumi Mochizuki, Dorota Wloga, François Juge, Krzysztof Rogowski
ABSTRACT
Tubulin posttranslational modifications represent an important mechanism involved in the regulation of microtubule functions. The most widespread among them are detyrosination, α∆2-tubulin, and polyglutamylation. Here, we describe a family of tubulin-modifying enzymes composed of two closely related proteins, KIAA0895L and KIAA0895, which have tubulin metallocarboxypeptidase activity and thus were termed TMCP1 and TMCP2, respectively. We show that TMCP1 (also known as MATCAP) acts as α-tubulin detyrosinase that also catalyzes α∆2-tubulin. In contrast, TMCP2 preferentially modifies βI-tubulin by removing three amino acids from its C terminus, generating previously unknown βI∆3 modification. We show that βI∆3-tubulin is mostly found on centrioles and mitotic spindles and in cilia. Moreover, we demonstrate that TMCPs also remove posttranslational polyglutamylation and thus act as tubulin deglutamylases. Together, our study describes the identification and comprehensive biochemical analysis of a previously unknown type of tubulin-modifying enzymes involved in the processing of α- and β-tubulin C-terminal tails and deglutamylation.
Sigma-Aldrich
Anti-β-Tubulin I antibody, Mouse monoclonal, clone SAP.4G5, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture