Merck
  • Early Events after Herpes Simplex Virus-Type 1 Entry Are Necessary for the Release of Gamma-Hydroxybutyrate upon Acute Infection.
Early Events after Herpes Simplex Virus-Type 1 Entry Are Necessary for the Release of Gamma-Hydroxybutyrate upon Acute Infection.
Pharmaceuticals (Basel, Switzerland) (2023-08-26)
Faith O Osinaga, Yu-Chih Chen, Madan K Kharel, Yan Waguespack, Sichu Li, Shaochung Victor Hsia
ABSTRACT
We reported that gamma-hydroxybutyrate (GHB) is released upon Herpes Simplex Virus Type-1 (HSV-1) acute infection. However, the cellular biochemical processes involved in the production of GHB in infected cells are unclear. This study aims to shed light on the biochemical pathway and the stage within the viral life cycle responsible for the release of GHB in infected cells. UV-inactivation, acyclovir (ACV), and cycloheximide (CHX) treatments were used to inhibit HSV-1 replication at various stages. Vero cells treated with UV-inactivated HSV-1 significantly decreased GHB production. However, ACV or CHX treatments did not affect GHB production. We also showed that inhibition of glycolytic enzyme enolase by sodium fluoride (NaF) significantly reduces GHB production upon infection. This finding suggests that suppression of glycolytic activity negatively affects cellular GHB production. Our data also indicated that succinic semialdehyde dehydrogenase, an enzyme involved in the shunt of the tricarboxylic acid (TCA) cycle to generate succinic acid, was decreased upon infection, suggesting that infection may trigger the accumulation of succinic semialdehyde, causing the production of GHB. Although the precise mechanism has yet to be defined, our results suggest that early events following infection modulates the release of GHB, which is generated through the metabolic pathways of glycolysis and TCA cycle.
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Pricing and availability is not currently available.