Merck
  • Transcriptional regulatory network during axonal regeneration of dorsal root ganglion neurons: laser-capture microdissection and deep sequencing.
Transcriptional regulatory network during axonal regeneration of dorsal root ganglion neurons: laser-capture microdissection and deep sequencing.
Neural regeneration research (2023-03-18)
Li-Li Zhao, Tao Zhang, Wei-Xiao Huang, Ting-Ting Guo, Xiao-Song Gu
ABSTRACT
The key regulators and regeneration-associated genes involved in axonal regeneration of neurons after injury have not been clarified. In high-throughput sequencing, various factors influence the final sequencing results, including the number and size of cells, the depth of sequencing, and the method of cell separation. There is still a lack of research on the detailed molecular expression profile during the regeneration of dorsal root ganglion neuron axon. In this study, we performed laser-capture microdissection coupled with RNA sequencing on dorsal root ganglion neurons at 0, 3, 6, and 12 hours and 1, 3, and 7 days after sciatic nerve crush in rats. We identified three stages after dorsal root ganglion injury: early (3-12 hours), pre-regeneration (1 day), and regeneration (3-7 days). Gene expression patterns and related function enrichment results showed that one module of genes was highly related to axonal regeneration. We verified the up-regulation of activating transcription factor 3 (Atf3), Kruppel like factor 6 (Klf6), AT-rich interaction domain 5A (Arid5a), CAMP responsive element modulator (Crem), and FOS like 1, AP-1 transcription factor Subunit (Fosl1) in dorsal root ganglion neurons after injury. Suppressing these transcription factors (Crem, Arid5a, Fosl1 and Klf6) reduced axonal regrowth in vitro. As the hub transcription factor, Atf3 showed higher expression and activity at the pre-regeneration and regeneration stages. G protein-coupled estrogen receptor 1 (Gper1), interleukin 12a (Il12a), estrogen receptor 1 (ESR1), and interleukin 6 (IL6) may be upstream factors that trigger the activation of Atf3 during the repair of axon injury in the early stage. Our study presents the detailed molecular expression profile during axonal regeneration of dorsal root ganglion neurons after peripheral nerve injury. These findings may provide reference for the clinical screening of molecular targets for the treatment of peripheral nerve injury.
Sigma-Aldrich
Anti-ARID5A (ab2) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Pricing and availability is not currently available.