Merck
  • Chemical pulldown combined with mass spectrometry to identify the molecular targets of antimalarials in cell-free lysates.
Chemical pulldown combined with mass spectrometry to identify the molecular targets of antimalarials in cell-free lysates.
STAR protocols (2023-01-08)
Robert J Smith, Rachel Milne, Victoriano Corpas Lopez, Natalie Wiedemar, Gourav Dey, Aisha J Syed, Stephen Patterson, Susan Wyllie
ABSTRACT
Here, we provide a protocol using chemical pulldown combined with mass spectrometry (LC-MS/MS) to identify drug targets in Plasmodium falciparum. This approach works upon the principle that a resin-bound inhibitor selectively binds its molecular target(s) in cell-free lysates. We describe the preparation of drug beads and P. falciparum lysate, followed by chemical pulldown, sample fractionation, and LC-MS/MS analysis. We then detail how to identify specifically bound proteins by comparing protein enrichment in DMSO-treated relative to drug-treated lysates via quantitative proteomics. For complete details on the use and execution of this protocol, please refer to Milne et al. (2022).1.
Sigma-Aldrich
Ethylenediaminetetraacetic acid tetrasodium salt hydrate, ≥99.0%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture