Merck
  • Chlorogenic acid and coffee prevent hypoxia-induced retinal degeneration.
Chlorogenic acid and coffee prevent hypoxia-induced retinal degeneration.
Journal of agricultural and food chemistry (2013-12-04)
Holim Jang, Hong Ryul Ahn, Hyoung Jo, Kyung-A Kim, Eun Ha Lee, Ki Won Lee, Sang Hoon Jung, Chang Y Lee
ABSTRACT
This study explored whether chlorogenic acid (CGA) and coffee have protective effects against retinal degeneration. Under hypoxic conditions, the viability of transformed retinal ganglion (RGC-5) cells was significantly reduced by treatment with the nitric oxide (NO) donor S-nitroso-N-acetylpenicillamine (SNAP). However, pretreatment with CGA attenuated cell death in a concentration-dependent manner. In addition, CGA prevented the up-regulation of apoptotic proteins such as Bad and cleaved caspase-3. Similar beneficial effects of both CGA and coffee extracts were observed in mice that had undergone an optic nerve crush (ONC) procedure. CGA and coffee extract reduced cell death by preventing the down-regulation of Thy-1. Our in vitro and in vivo studies demonstrated that coffee and its major component, CGA, significantly reduce apoptosis of retinal cells induced by hypoxia and NO, and that coffee consumption may help in preventing retinal degeneration.
Sigma-Aldrich
3-O-p-Coumaroylquinic acid, ≥95%
Sigma-Aldrich
5-O-p-Coumaroylquinic acid, ≥95%
Sigma-Aldrich
4-O-p-Coumaroylquinic acid