Merck
  • Histone methyltransferase SUV39H1 regulates the Golgi complex via the nuclear envelope-spanning LINC complex.
Histone methyltransferase SUV39H1 regulates the Golgi complex via the nuclear envelope-spanning LINC complex.
PloS one (2023-07-12)
Miyu Nishino, Hiromasa Imaizumi, Yuhki Yokoyama, Jun Katahira, Hiroshi Kimura, Nariaki Matsuura, Miki Matsumura
ABSTRACT
Cell motility is related to the higher-order structure of chromatin. Stimuli that induce cell migration change chromatin organization; such stimuli include elevated histone H3 lysine 9 trimethylation (H3K9me3). We previously showed that depletion of histone H3 lysine 9 methyltransferase, SUV39H1, suppresses directional cell migration. However, the molecular mechanism underlying this association between chromatin and cell migration remains elusive. The Golgi apparatus is a cell organelle essential for cell motility. In this study, we show that loss of H3K9 methyltransferase SUV39H1 but not SETDB1 or SETDB2 causes dispersion of the Golgi apparatus throughout the cytoplasm. The Golgi dispersion triggered by SUV39H1 depletion is independent of transcription, centrosomes, and microtubule organization, but is suppressed by depletion of any of the following three proteins: LINC complex components SUN2, nesprin-2, or microtubule plus-end-directed kinesin-like protein KIF20A. In addition, SUN2 is closely localized to H3K9me3, and SUV39H1 affects the mobility of SUN2 in the nuclear envelope. Further, inhibition of cell motility caused by SUV39H1 depletion is restored by suppression of SUN2, nesprin-2, or KIF20A. In summary, these results show the functional association between chromatin organization and cell motility via the Golgi organization regulated by the LINC complex.
Sigma-Aldrich
ANTI-SUN1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-SUV39H1 Antibody, clone MG44, ascites fluid, clone MG44, Upstate®
Sigma-Aldrich
Anti-Golgi FTCD antibody, clone 58k-9, Mouse monoclonal, ascites fluid
Sigma-Aldrich
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution