Merck
  • Monoacidic Inhibitors of the Kelch-like ECH-Associated Protein 1: Nuclear Factor Erythroid 2-Related Factor 2 (KEAP1:NRF2) Protein-Protein Interaction with High Cell Potency Identified by Fragment-Based Discovery.
Monoacidic Inhibitors of the Kelch-like ECH-Associated Protein 1: Nuclear Factor Erythroid 2-Related Factor 2 (KEAP1:NRF2) Protein-Protein Interaction with High Cell Potency Identified by Fragment-Based Discovery.
Journal of medicinal chemistry (2016-04-01)
Thomas G Davies, William E Wixted, Joseph E Coyle, Charlotte Griffiths-Jones, Keisha Hearn, Rachel McMenamin, David Norton, Sharna J Rich, Caroline Richardson, Gordon Saxty, Henriëtte M G Willems, Alison J-A Woolford, Joshua E Cottom, Jen-Pyng Kou, John G Yonchuk, Heidi G Feldser, Yolanda Sanchez, Joseph P Foley, Brian J Bolognese, Gregory Logan, Patricia L Podolin, Hongxing Yan, James F Callahan, Tom D Heightman, Jeffrey K Kerns
ABSTRACT
KEAP1 is the key regulator of the NRF2-mediated cytoprotective response, and increasingly recognized as a target for diseases involving oxidative stress. Pharmacological intervention has focused on molecules that decrease NRF2-ubiquitination through covalent modification of KEAP1 cysteine residues, but such electrophilic compounds lack selectivity and may be associated with off-target toxicity. We report here the first use of a fragment-based approach to directly target the KEAP1 Kelch-NRF2 interaction. X-ray crystallographic screening identified three distinct "hot-spots" for fragment binding within the NRF2 binding pocket of KEAP1, allowing progression of a weak fragment hit to molecules with nanomolar affinity for KEAP1 while maintaining drug-like properties. This work resulted in a promising lead compound which exhibits tight and selective binding to KEAP1, and activates the NRF2 antioxidant response in cellular and in vivo models, thereby providing a high quality chemical probe to explore the therapeutic potential of disrupting the KEAP1-NRF2 interaction.
Sigma-Aldrich
KI-696, ≥98% (HPLC)