Merck
  • The alteration of intestinal mucosal α-synuclein expression and mucosal microbiota in Parkinson's disease.
The alteration of intestinal mucosal α-synuclein expression and mucosal microbiota in Parkinson's disease.
Applied microbiology and biotechnology (2023-02-17)
Jihua Shi, Yiran Wang, Dan Chen, Xue Xu, Wenbin Li, Kai Li, Jing He, Wen Su, Qingfeng Luo
ABSTRACT
Parkinson's disease (PD) is the second most common neurodegenerative disease but still lacks a preclinical strategy to identify it. The diagnostic value of intestinal mucosal α-synuclein (αSyn) in PD has not drawn a uniform conclusion. The relationship between the alteration of intestinal mucosal αSyn expression and mucosal microbiota is unclear. Nineteen PD patients and twenty-two healthy controls were enrolled in our study from whom were collected, using gastrointestinal endoscopes, duodenal and sigmoid mucosal samples for biopsy. Multiplex immunohistochemistry was performed to detect total, phosphorylate, and oligomer α-synuclein. Next-generation 16S rRNA amplicon sequencing was applied for taxonomic analysis. The results implied that oligomer α-synuclein (OSyn) in sigmoid mucosa of PD patients was transferred from the intestinal epithelial cell membrane to the cytoplasm, acinar lumen, and stroma. Its distribution feature was significantly different between the two groups, especially the ratio of OSyn/αSyn. The microbiota composition in mucosa also differed. The relative abundances of Kiloniellales, Flavobacteriaceae, and CAG56 were lower, while those of Proteobacteria, Gammaproteobacteria, Burkholderiales, Burkholdriaceae, Oxalobacteraceae, Ralstonia, Massilla, and Lactoccus were higher in duodenal mucosa of PD patients. The relative abundances of Thermoactinomycetales and Thermoactinomycetaceae were lower, while those of Prevotellaceae and Bifidobacterium longum were higher in patients' sigmoid mucosa. Further, the OSyn/αSyn level was positively correlated with the relative abundances of Proteobacteria, Gammaproteobacteria, Burkholderiales, Pseudomonadales, Burkholderiaceae, and Ralstonia in the duodenal mucosa, while it was negatively correlated with the Chao1 index and observed operational taxonomic units of microbiota in sigmoid mucosa. The intestinal mucosal microbiota composition of PD patients altered with the relative abundances of proinflammatory bacteria in the duodenal mucosa increased. The ratio of the OSyn/αSyn level in the sigmoid mucosa indicated a potential diagnostic value for PD, which also correlated with mucosal microbiota diversity and composition. KEY POINTS: • The distribution of OSyn in sigmoid mucosa differed between PD patients and healthy controls. • Significant alterations in the microbiome were found in PD patients' gut mucosa. • OSyn/αSyn level in sigmoid mucosa indicated a potential diagnostic value for PD.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-alpha-Synuclein Antibody, oligomer-specific Syn33, from rabbit
Sigma-Aldrich
Anti-phospho-α-Synuclein (Ser129) Antibody, clone 81A, clone 81A, from mouse