Merck
  • Photoreceptor laminin drives differentiation of human pluripotent stem cells to photoreceptor progenitors that partially restore retina function.
Photoreceptor laminin drives differentiation of human pluripotent stem cells to photoreceptor progenitors that partially restore retina function.
Molecular therapy : the journal of the American Society of Gene Therapy (2023-01-14)
Hwee Goon Tay, Helder Andre, Vicki Chrysostomou, Swarnaseetha Adusumalli, Jing Guo, Xiaoyuan Ren, Wei Sheng Tan, Jia En Tor, Aida Moreno-Moral, Flavia Plastino, Hammurabi Bartuma, Zuhua Cai, Sai Bo Bo Tun, Veluchamy Amutha Barathi, Gavin Tan Siew Wei, Gianluca Grenci, Li Yen Chong, Arne Holmgren, Anders Kvanta, Crowston Jonathan Guy, Enrico Petretto, Karl Tryggvason
ABSTRACT
Blindness caused by advanced stages of inherited retinal diseases and age-related macular degeneration are characterized by photoreceptor loss. Cell therapy involving replacement with functional photoreceptor-like cells generated from human pluripotent stem cells holds great promise. Here, we generated a human recombinant retina-specific laminin isoform, LN523, and demonstrated the role in promoting the differentiation of human embryonic stem cells into photoreceptor progenitors. This chemically defined and xenogen-free method enables reproducible production of photoreceptor progenitors within 32 days. We observed that the transplantation into rd10 mice were able to protect the host photoreceptor outer nuclear layer (ONL) up to 2 weeks post transplantation as measured by full-field electroretinogram. At 4 weeks post transplantation, the engrafted cells were found to survive, mature, and associate with the host's rod bipolar cells. Visual behavioral assessment using the water maze swimming test demonstrated visual improvement in the cell-transplanted rodents. At 20 weeks post transplantation, the maturing engrafted cells were able to replace the loss of host ONL by extensive association with host bipolar cells and synapses. Post-transplanted rabbit model also provided congruent evidence for synaptic connectivity with the degenerated host retina. The results may pave the way for the development of stem cell-based therapeutics for retina degeneration.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-Goat IgG (H+L), highly cross-adsorbed, CF 647 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit
Sigma-Aldrich
CKI-7 dihydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Anti-LAMC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution