Merck
  • Targeting MYO1B impairs tumorigenesis via inhibiting the SNAI2/cyclin D1 signaling in esophageal squamous cell carcinoma.
Targeting MYO1B impairs tumorigenesis via inhibiting the SNAI2/cyclin D1 signaling in esophageal squamous cell carcinoma.
Journal of cellular physiology (2022-07-22)
Shau-Hsuan Li, Limei Qian, Yen-Hao Chen, Hung-I Lu, Hsin-Ting Tsai, Haiwen Lyu, Runxiang Yang, Chang-Han Chen
ABSTRACT
Myosin-related proteins play an important role in cancer progression. However, the clinical significance, biological functions, and mechanisms of myosin 1B (MYO1B), in esophageal squamous cell carcinoma (ESCC) remain unclear. The clinical relevance of MYO1B, SNAI2, and cyclin D1 in ESCC was determined by immunohistochemistry, Oncomine, and GEPIA databases. The oncogenic roles of MYO1B were determined by CCK8, colony formation assays, wound healing, and Transwell assay. MYO1B, SNAI2, and cyclin D1 at mRNA and protein levels in ESCC cells were detected by qPCR and Western blot analysis. In our study, we found that MYO1B expression was increased in ESCC tissue samples and correlated with tumor stage, TNM stage, and poor outcomes. Functional assays indicated that depletion of MYO1B impaired oncogenesis, and enhanced chemosensitivity in ESCC. Bioinformatic analysis and mechanistic studies illustrated that SNAI2 was a key downstream effector of MYO1B. Suppression of MYO1B downregulated expression of SNAI2, thereby inhibiting the SNAI2/cyclin D1 pathway. Furthermore, a selective inhibitor of cyclin D1 activation reversed siMYO1B cells overexpressing SNAI2-elicited aggressive phenotypes of ESCC cells. MYO1B positively correlated with SNAI2 and cyclin D1 in ESCC samples, and higher SNAI2 expression was also associated with poor prognosis in ESCC patients. Our finding demonstrated that MYO1B activates the SNAI2/cyclin D1 pathway to drive tumorigenesis and cisplatin cytotoxicity in ESCC, indicating that MYO1B is a potential therapeutic target for patients with ESCC.
Sigma-Aldrich
Anti-MYO1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution