- Generation and Functional Characterization of PLAP CAR-T Cells against Cervical Cancer Cells.
Generation and Functional Characterization of PLAP CAR-T Cells against Cervical Cancer Cells.
Chimeric antigen receptor (CAR) T-cell therapy is one of the cancer treatment modalities that has recently shown promising results in treating hematopoietic malignancies. However, one of the obstacles that need to be addressed in solid tumors is the on-target and off-tumor cytotoxicity due to the lack of specific tumor antigens with low expression in healthy cells. Placental alkaline phosphatase (PLAP) is a shared placenta- and tumor-associated antigen (TAA) that is expressed in ovarian, cervical, colorectal, and prostate cancers and is negligible in normal cells. In this study, we constructed second-generation CAR T cells with a fully human scFv against PLAP antigen andthen evaluated the characteristics of PLAP CAR T cells in terms of tonic signaling and differentiation in comparison with ΔPLAP CAR T cells and CD19 CAR T cells. In addition, by co-culturing PLAP CAR T cells with HeLa and CaSki cells, we analyzed the tumor-killing functions and the secretion of anti-tumor molecules. Results showed that PLAP CAR T cells not only proliferated during co-culture with cancer cells but also eliminated them in vitro. We also observed increased secretion of IL-2, granzyme A, and IFN-γ by PLAP CAR T cells upon exposure to the target cells. In conclusion, PLAP CAR T cells are potential candidates for further investigation in cervical cancer and, potentially, other solid tumors.