Merck
  • The V2 receptor antagonist tolvaptan counteracts proliferation and invasivity in human cancer cells.
The V2 receptor antagonist tolvaptan counteracts proliferation and invasivity in human cancer cells.
Journal of endocrinological investigation (2022-05-24)
G Marroncini, C Anceschi, L Naldi, B Fibbi, F Baldanzi, M Maggi, A Peri
ABSTRACT
Hyponatremia, the most frequent electrolyte alteration in clinical practice, has been associated with a worse prognosis in cancer patients. On the other hand, a better outcome has been related to serum sodium normalization. In vitro studies have shown that low extracellular sodium promotes cancer cells proliferation and invasiveness. Tolvaptan, a selective vasopressin receptor type 2 (V2) antagonist, has been effectively used in the last decade for the treatment of hyponatremia secondary to the Syndrome of Inappropriate Antidiuresis. A few in vitro data suggested a direct role of tolvaptan in counteracting cancer progression, so far. The aim of this study was to evaluate the effect and the mechanism of action of tolvaptan in cell lines from different tumours [i.e. colon cancer (HCT-8), hepatocarcinoma (HepG2), neuroblastoma (SK-N-AS)]. First, we showed that these cell lines express the V2 receptor. Tolvaptan significantly reduced cell proliferation with an IC50 in the micromolar range. Accordingly, reduced levels of cAMP, of the catalytic α subunit of PKA, and a reduced pAKT/AKT ratio were observed. Tolvaptan effectively inhibited cell cycle progression, whereas it induced apoptotis. Furthermore, it reduced cell invasiveness. In particular, anchorage-independent growth and the activity of collagenases type IV were blunted in the three cell lines. Accordingly, tolvaptan counteracted the RhoA/ROCK1-2 pathway, which has a pivotal role in regulating cell movement. Overall, these findings indicate that tolvaptan effectively inhibits tumour progression in vitro. Further studies should clarify whether the V2 receptor might be considered a possible target in anti-cancer strategies in the future.
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-phospho-Akt (Ser473) Antibody, clone 6F5, clone 6F5, from mouse
Pricing and availability is not currently available.