Merck
  • Impact of immobilization strategies on the activity and recyclability of lipases in nanomagnetic supports.
Impact of immobilization strategies on the activity and recyclability of lipases in nanomagnetic supports.
Scientific reports (2022-04-28)
Thais de Andrade Silva, Wanderson Juvêncio Keijok, Marco Cesar Cunegundes Guimarães, Sérvio Túlio Alves Cassini, Jairo Pinto de Oliveira
ABSTRACT
The use of enzymes immobilized on nanomagnetic supports has produced surprising results in catalysis, mainly due to the increase in surface area and the potential for recovery and reuse. However, the meticulous control of the process and difficulties in reproducibility have made industrial-scale applications unfeasible. Furthermore, the role of conjugation strategies in the catalytic activity and recycling of catalysts is unclear. Therefore, the objective of this study was to compare the conjugation of enzymes on nanomagnetic supports through physical adsorption (naked) or covalent bonding with mercaptopropyltrimethoxysilane (MPTS) and aminopropyltriethoxysilane (APTS) ligands. The free lipase obtained from Rhizomucor miehei was used as a model enzyme. Total protein and enzyme activity were determined using spectrophotometry (UV-Vis) and the p-nitrophenyl palmitate (p-NPP) hydrolysis method. The results indicated that a more significant enzyme surface loading does not always mean better immobilization success. The physical adsorption binding strategy had higher surface loading and low catalytic activity. On the other hand, covalent coupling with free NH2 had an excellent catalytic activity with very low surface loading. Finally, we show that recyclability can be improved with conjugation mediated by disulfide bonds. The findings presented here are essential for developing nanoconjugates with high enzymatic activity, which can guarantee the success of several industrial applications.
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Pricing and availability is not currently available.