Merck
  • Transformation of human umbilical mesenchymal cells into neurons in vitro.
Transformation of human umbilical mesenchymal cells into neurons in vitro.
Journal of biomedical science (2004-08-19)
Yu-Show Fu, Yu-Tsung Shih, Yun-Chih Cheng, Ming-Yuan Min
ABSTRACT
Neuronal transplantation has provided a promising approach for treating neurodegenerative diseases. Recently, efforts have been directed at in vitro induction of various stem cells to transform into neurons. We report the first successful quantities in an in vitro attempt at directing the transformation into neurons of human umbilical mesenchymal cells, which are capable of rapid proliferation in vitro and are easily available. When cultured in neuronal conditioned medium, human umbilical mesenchymal cells started to express neuron-specific proteins such as NeuN and neurofilament (NF) on the 3rd day and exhibited retraction of the cell body, elaboration of processes, clustering of cells and expression of functional mRNA responsible for the synthesis of subunits of the kainate receptor and glutamate decarboxylase on the 6th day. Between the 9th and 12th days, the percentage of human umbilical mesenchymal cells expressing NF was as high as 87%, while functionality was demonstrated by glutamate invoking an inward current. At this stage, cells were differentiated into mature neurons in the post mitosis phase.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) FITC Conjugated, 2 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-BrdU Antibody, clone IIB5, clone IIB5, Chemicon®, from mouse
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-KA2/GRIK5 (Kainate Receptor) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse