Merck
  • CD4+ T-cell-derived IL-10 promotes CNS inflammation in mice by sustaining effector T cell survival.
CD4+ T-cell-derived IL-10 promotes CNS inflammation in mice by sustaining effector T cell survival.
Cell reports (2022-03-31)
Nir Yogev, Tanja Bedke, Yasushi Kobayashi, Leonie Brockmann, Dominika Lukas, Tommy Regen, Andrew L Croxford, Alexei Nikolav, Nadine Hövelmeyer, Esther von Stebut, Marco Prinz, Carles Ubeda, Kevin J Maloy, Nicola Gagliani, Richard A Flavell, Ari Waisman, Samuel Huber
ABSTRACT
Interleukin (IL)-10 is considered a prototypical anti-inflammatory cytokine, significantly contributing to the maintenance and reestablishment of immune homeostasis. Accordingly, it has been shown in the intestine that IL-10 produced by Tregs can act on effector T cells, thereby limiting inflammation. Herein, we investigate whether this role also applies to IL-10 produced by T cells during central nervous system (CNS) inflammation. During neuroinflammation, both CNS-resident and -infiltrating cells produce IL-10; yet, as IL-10 has a pleotropic function, the exact contribution of the different cellular sources is not fully understood. We find that T-cell-derived IL-10, but not other relevant IL-10 sources, can promote inflammation in experimental autoimmune encephalomyelitis. Furthermore, in the CNS, T-cell-derived IL-10 acts on effector T cells, promoting their survival and thereby enhancing inflammation and CNS autoimmunity. Our data indicate a pro-inflammatory role of T-cell-derived IL-10 in the CNS.
Sigma-Aldrich
Avidin–Peroxidase, buffered aqueous solution
Pricing and availability is not currently available.