Merck
  • Tankyrase represses autoinflammation through the attenuation of TLR2 signaling.
Tankyrase represses autoinflammation through the attenuation of TLR2 signaling.
The Journal of clinical investigation (2022-04-02)
Yoshinori Matsumoto, Ioannis D Dimitriou, Jose La Rose, Melissa Lim, Susan Camilleri, Napoleon Law, Hibret A Adissu, Jiefei Tong, Michael F Moran, Andrzej Chruscinski, Fang He, Yosuke Asano, Takayuki Katsuyama, Ken-Ei Sada, Jun Wada, Robert Rottapel
ABSTRACT
Dysregulation of Toll-like receptor (TLR) signaling contributes to the pathogenesis of autoimmune diseases. Here, we provide genetic evidence that tankyrase, a member of the poly(ADP-ribose) polymerase (PARP) family, negatively regulates TLR2 signaling. We show that mice lacking tankyrase in myeloid cells developed severe systemic inflammation with high serum inflammatory cytokine levels. We provide mechanistic evidence that tankyrase deficiency resulted in tyrosine phosphorylation and activation of TLR2 and show that phosphorylation of tyrosine 647 within the TIR domain by SRC and SYK kinases was critical for TLR2 stabilization and signaling. Last, we show that the elevated cytokine production and inflammation observed in mice lacking tankyrase in myeloid cells were dependent on the adaptor protein 3BP2, which is required for SRC and SYK activation. These data demonstrate that tankyrase provides a checkpoint on the TLR-mediated innate immune response.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-MyD88 antibody produced in mouse, clone 2E9C2, purified immunoglobulin, buffered aqueous solution