Merck
A semi-dominant NLR allele causes whole-seedling necrosis in wheat.
Plant physiology (2021-02-13)
Haiyan Jia, Shulin Xue, Lei Lei, Min Fan, Shuxia Peng, Tian Li, Ragupathi Nagarajan, Brett Carver, Zhengqiang Ma, Junpeng Deng, Liuling Yan
ABSTRACT
Programmed cell death (PCD) and apoptosis have key functions in development and disease resistance in diverse organisms; however, the induction of necrosis remains poorly understood. Here, we identified a semi-dominant mutant allele that causes the necrotic death of the entire seedling (DES) of wheat (Triticum aestivum L.) in the absence of any pathogen or external stimulus. Positional cloning of the lethal allele mDES1 revealed that this premature death via necrosis was caused by a point mutation from Asp to Asn at amino acid 441 in a nucleotide-binding leucine-rich repeat protein containing nucleotide-binding domain and leucine-rich repeats. The overexpression of mDES1 triggered necrosis and PCD in transgenic plants. However, transgenic wheat harboring truncated wild-type DES1 proteins produced through gene editing that exhibited no significant developmental defects. The point mutation in mDES1 did not cause changes in this protein in the oligomeric state, but mDES1 failed to interact with replication protein A leading to abnormal mitotic cell division. DES1 is an ortholog of Sr35, which recognizes a Puccinia graminis f. sp. tritici stem rust disease effector in wheat, but mDES1 gained function as a direct inducer of plant death. These findings shed light on the intersection of necrosis, apoptosis, and autoimmunity in plants.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-6X His IgG, CF 488A antibody produced in mouse, ~2 mg/mL, clone 33D10.D2.G8, purified immunoglobulin