Merck
  • Gene transfer of tissue inhibitor of metalloproteinases-3 reverses the inhibitory effects of TNF-alpha on Fas-induced apoptosis in rheumatoid arthritis synovial fibroblasts.
Gene transfer of tissue inhibitor of metalloproteinases-3 reverses the inhibitory effects of TNF-alpha on Fas-induced apoptosis in rheumatoid arthritis synovial fibroblasts.
Journal of immunology (Baltimore, Md. : 1950) (2005-05-10)
Andreas Drynda, Paul H A Quax, Manfred Neumann, Willemijn H van der Laan, Géza Pap, Susanne Drynda, Ingmar Meinecke, Joern Kekow, Wolfram Neumann, Tom W J Huizinga, Michael Naumann, Wolfgang König, Thomas Pap
ABSTRACT
Apart from counteracting matrix metalloproteinases, tissue inhibitor of metalloproteinases-3 (TIMP-3) has proapoptotic properties. These features have been attributed to the inhibition of metalloproteinases involved in the shedding of cell surface receptors such as the TNFR. However, little is known about effects of TIMP-3 in cells that are not susceptible to apoptosis by TNF-alpha. In this study, we report that gene transfer of TIMP-3 into human rheumatoid arthritis synovial fibroblasts and MRC-5 human fetal lung fibroblasts facilitates apoptosis and completely reverses the apoptosis-inhibiting effects of TNF-alpha. Although TNF-alpha inhibits Fas/CD95-induced apoptosis in untransfected and mock-transfected cells, fibroblasts ectopically expressing TIMP-3 are sensitized most strongly to Fas/CD95-mediated cell death by TNF-alpha. Neither synthetic MMP inhibitors nor glycosylated bioactive TIMP-3 are able to achieve these effects. Gene transfer of TIMP-3 inhibits the TNF-alpha-induced activation of NF-kappaB in rheumatoid arthritis synovial fibroblasts and reduces the up-regulation of soluble Fas/CD95 by TNF-alpha, but has no effects on the cell surface expression of Fas. Collectively, our data demonstrate that intracellularly produced TIMP-3 not only induces apoptosis, but also modulates the apoptosis-inhibiting effects of TNF-alpha in human rheumatoid arthritis synovial fibroblast-like cells. Thus, our findings may stimulate further studies on the therapeutic potential of gene transfer strategies with TIMP-3.
Sigma-Aldrich
Anti-NFκB p52 Antibody, Upstate®, from rabbit