Merck
  • The PGRMC1 Antagonist AG-205 Inhibits Synthesis of Galactosylceramide and Sulfatide.
The PGRMC1 Antagonist AG-205 Inhibits Synthesis of Galactosylceramide and Sulfatide.
Cells (2021-12-25)
Lihua Wang-Eckhardt, Ivonne Becker, Matthias Eckhardt
ABSTRACT
Sulfatide synthesis in the human renal cancer cell line SMKT-R3 was strongly inhibited in the presence of low µM concentrations of AG-205, a progesterone receptor membrane component 1 (PGRMC1) antagonist. This was also the case in Chinese hamster ovary (CHO) cells stably transfected with UDP-galactose: ceramide galactosyltransferase and cerebroside sulfotransferase, the two enzymes required for sulfatide synthesis. In CHO cells synthesizing galactosylceramide but not sulfatide, galactosylceramide was also strongly reduced, suggesting an effect at the level of galactolipid synthesis. Notably, AG-205 inhibited galactosylceramide synthesis to a similar extent in wild type CHO cells and cells that lack PGRMC1 and/or PGRMC2. In vitro enzyme activity assays showed that AG-205 is an inhibitor of UDP-galactose: ceramide galactosyltransferase, but not cerebroside sulfotransferase. This study shows that PGRMC1 is only one of several targets of AG-205 and should be used with caution, especially in studies using cells synthesizing galactosylceramide and sulfatide.
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC), Molecular Biology
Sigma-Aldrich
G 418 disulfate salt solution, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
4-Methylumbelliferyl sulfate potassium salt, sulfatase substrate