Merck
  • Ankmy2 Prevents Smoothened-Independent Hyperactivation of the Hedgehog Pathway via Cilia-Regulated Adenylyl Cyclase Signaling.
Ankmy2 Prevents Smoothened-Independent Hyperactivation of the Hedgehog Pathway via Cilia-Regulated Adenylyl Cyclase Signaling.
Developmental cell (2020-07-24)
Bandarigoda Nipunika Somatilaka, Sun-Hee Hwang, Vivek Reddy Palicharla, Kevin Andrew White, Hemant Badgandi, John Michael Shelton, Saikat Mukhopadhyay
ABSTRACT
The mechanisms underlying subcellular targeting of cAMP-generating adenylyl cyclases and processes regulated by their compartmentalization are poorly understood. Here, we identify Ankmy2 as a repressor of the Hedgehog pathway via adenylyl cyclase targeting. Ankmy2 binds to multiple adenylyl cyclases, determining their maturation and trafficking to primary cilia. Mice lacking Ankmy2 are mid-embryonic lethal. Knockout embryos have increased Hedgehog signaling and completely open neural tubes showing co-expansion of all ventral neuroprogenitor markers, comparable to the loss of the Hedgehog receptor Patched1. Ventralization in Ankmy2 knockout is completely independent of the Hedgehog pathway transducer Smoothened. Instead, ventralization results from the reduced formation of Gli2 and Gli3 repressors and early depletion of adenylyl cyclase III in neuroepithelial cilia, implicating deficient pathway repression. Ventralization in Ankmy2 knockout requires both cilia and Gli2 activation. These findings indicate that cilia-dependent adenylyl cyclase signaling represses the Hedgehog pathway and promotes morphogenetic patterning.
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-S•Tag fusion protein Antibody, clone 26A4.1.2, from mouse
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
SYBR® Green Quantitative RT-qPCR Kit, One step SYBR® Green RT-qPCR with MMLV & hot-start Taq DNA Polymerase
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
Anti-ANKMY2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody