Merck
  • Tissue differences in the exosomal/small extracellular vesicle proteome and their potential as indicators of altered tissue metabolism.
Tissue differences in the exosomal/small extracellular vesicle proteome and their potential as indicators of altered tissue metabolism.
Cell reports (2022-01-20)
Ruben Garcia-Martin, Bruna Brasil Brandao, Thomas Thomou, Emrah Altindis, C Ronald Kahn
ABSTRACT
Exosomes/small extracellular vesicles (sEVs) can serve as multifactorial mediators of cell-to-cell communication through their miRNA and protein cargo. Quantitative proteomic analysis of five cell lines representing metabolically important tissues reveals that each cell type has a unique sEV proteome. While classical sEV markers such as CD9/CD63/CD81 vary markedly in abundance, we identify six sEV markers (ENO1, GPI, HSPA5, YWHAB, CSF1R, and CNTN1) that are similarly abundant in sEVs of all cell types. In addition, each cell type has specific sEV markers. Using fat-specific Dicer-knockout mice with decreased white adipose tissue and increased brown adipose tissue, we show that these cell-type-specific markers can predict the changing origin of the serum sEVs. These results provide a valuable resource for understanding the sEV proteome of the cells and tissues important in metabolic homeostasis, identify unique sEV markers, and demonstrate how these markers can help in predicting the tissue of origin of serum sEVs.
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Adiponectin Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Pricing and availability is not currently available.
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Pricing and availability is not currently available.