Merck
  • SARS-CoV-2 protein subunit vaccination of mice and rhesus macaques elicits potent and durable neutralizing antibody responses.
SARS-CoV-2 protein subunit vaccination of mice and rhesus macaques elicits potent and durable neutralizing antibody responses.
Cell reports. Medicine (2021-04-13)
Marco Mandolesi, Daniel J Sheward, Leo Hanke, Junjie Ma, Pradeepa Pushparaj, Laura Perez Vidakovics, Changil Kim, Monika Àdori, Klara Lenart, Karin Loré, Xaquin Castro Dopico, Jonathan M Coquet, Gerald M McInerney, Gunilla B Karlsson Hedestam, Ben Murrell
ABSTRACT
The outbreak and spread of SARS-CoV-2 (severe acute respiratory syndrome-coronavirus-2) is a current global health emergency, and effective prophylactic vaccines are needed urgently. The spike glycoprotein of SARS-CoV-2 mediates entry into host cells, and thus is the target of neutralizing antibodies. Here, we show that adjuvanted protein immunization with soluble SARS-CoV-2 spike trimers, stabilized in prefusion conformation, results in potent antibody responses in mice and rhesus macaques, with neutralizing antibody titers exceeding those typically measured in SARS-CoV-2 seropositive humans by more than one order of magnitude. Neutralizing antibody responses were observed after a single dose, with exceptionally high titers achieved after boosting. A follow-up to monitor the waning of the neutralizing antibody responses in rhesus macaques demonstrated durable responses that were maintained at high and stable levels at least 4 months after boosting. These data support the development of adjuvanted SARS-CoV-2 prefusion-stabilized spike protein subunit vaccines.
Sigma-Aldrich
Staphylococcal enterotoxin B from Staphylococcus aureus
Sigma-Aldrich
SIGMAFAST OPD, tablet