Merck
  • Tyrosine sulfation and O-glycosylation of chemoattractant receptor GPR15 differentially regulate interaction with GPR15L.
Tyrosine sulfation and O-glycosylation of chemoattractant receptor GPR15 differentially regulate interaction with GPR15L.
Journal of cell science (2021-03-25)
Yukari Okamoto, Sojin Shikano
ABSTRACT
GPR15 is a G-protein-coupled receptor (GPCR) that directs lymphocyte homing to the colon and skin. Recent studies have identified a chemokine-like protein GPR15L (also known as C10orf99) as a functional ligand of GPR15. In this study, we examined the structural elements that regulate the GPR15-GPR15L interaction with primary focus on post-translational modifications (PTMs) of receptor N-terminus and on the C-terminus of the ligand. Our findings reveal that the GPR15 receptor is sulfated on the N-terminal tyrosine residue(s) and disruption of tyrosine sulfation inhibits binding of GPR15L. In contrast, the disruption of O-glycosylation on the N-terminal threonine or serine residues, or the removal of α2,3-linked sialic acids from O-glycans, enhances the GPR15L binding. Thus, GPR15 represents a unique chemoattractant receptor in which different N-terminal PTMs regulate its ligand binding in a contrasting manner. We further demonstrate that, unlike canonical chemokines, GPR15L activity critically requires its extreme C-terminal residue and that its hydrophobicity may be a key attribute that facilitates an optimal interaction with the receptor. Our results reveal novel insights into chemoattractant receptor-ligand interaction and provide a valid footing for potential intervention targeting the GPR15-GPR15L axis.
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, Alexa Fluor 488 conjugate, clone 4A6, Upstate®, from mouse